Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
U heeft gezocht op: "rna-zuiveringskits"
https://nl.vwr.com/stibo/smallweb/std.lang.all/96/97/18989697.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"51"
  List view Grid view Easy View (new)
Sorteer op:
Leverancier:  VWR Chemicals
Omschrijving:   E.Z.N.A.® Plant RNA Kit provides a convenient and rapid method for the isolation of total RNA from a variety of plant samples. This kit provides a hom...
Leverancier:  VWR Chemicals
Omschrijving:   De E.Z.N.A.® MicroElute Total RNA-kit biedt een snelle en eenvoudige methode voor het isoleren van maximaal 50 µg totaal RNA uit kleine hoeveelheden g...
Leverancier:  VWR Chemicals
Omschrijving:   The E.Z.N.A.® Soil RNA Kit is designed to isolate high quality of total RNA from soil samples typically containing humic acid and inhibitor of RT-PCR....
Catalogus nummer: (R6732-00)
Leverancier:  VWR Chemicals
Omschrijving:   Zuivering van nucleïnezuren, RNA-zuiveringskit, E-Z 96®, E-Z 96® Total DNA/RNA Isolatie Kit (1×96)
UOM:  1 * 1 EA
Leverancier:  VWR Chemicals
Omschrijving:   Zuivering van nucleïnezuren, RNA-zuiveringskit, E-Z 96®, E-Z 96® Total RNA-kit II (4×96)
Leverancier:  VWR Chemicals
Omschrijving:   The Mag-Bind® Total RNA Kit allows the rapid and reliable isolation of high-quality total cellular RNA from a wide variety of cells and tissues. The s...
Leverancier:  VWR Chemicals
Omschrijving:   E.Z.N.A.® Tissue RNA Kit provides a rapid and easy method for the isolation of total RNA from difficult-to-lyse animal tissue samples including skelet...
Leverancier:  VWR Chemicals
Omschrijving:   The Mag-Bind® PX Blood RNA Kit allows for the isolation of total RNA from 2,5 ml of whole blood that has been stored in PAXgene® Blood RNA tubes. Stab...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 51
  1  2  3  4  5  6  7  Volgende