Pallets and sumps for storage of hazardous materials
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA18451EA 1480 EUR
MSGA18451 MSGA18450
Pallets and sumps for storage of hazardous materials
PE, blue. Safe storage of aggressive liquids such as acids and alkalis.
  • For internal and external use
  • Depot manufactured completely from polyethylene
  • Roller slide front closing door
  • Integrated sump
  • Combined storing of drums and small containers is possible
Opgelet: Not suitable for the storage of flammable liquids.