trans-2-Hexenyl acetate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2497-18-9
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00009474
:142.20 g/mol
Boiling Pt:165°C
Flash Pt:64°C
EINECS:219-680-7
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: A0877-25MLLocal Article Number: TCIAA0877-25ML
- Supplier Article Number: A0877-250MLLocal Article Number: TCIAA0877-250ML
Frequently Bought Together