HPLC columns, AquaSil™ C18
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AquaSil™
HYPE77503-153030EA 1110 EUR
HYPE77503-153030 HYPE77505-124030 HYPE77505-254030 HYPE77503-103030 HYPE77505-254630 HYPE77505-153030 HYPE77503-014001 HYPE77503-012101 HYPE77505-012101 HYPE77505-014001 HYPE77505-034030 HYPE77505-102130 HYPE77503-154030 HYPE77505-104630 HYPE77505-152130 HYPE77503-152130 HYPE77505-253030 HYPE77503-102130 HYPE77503-154630 HYPE77505-154630 HYPE77503-104630 HYPE77505-154030 HYPE77503-013001 HYPE77505-013001 HYPE77503-054630 HYPE77505-054630 HYPE77503-052130 HYPE77505-052130
HPLC columns, AquaSil™ C18
Kolommen voor chromatografie
The AquaSil™ C18 columns provide a versatile C18 phase for a wide range of applications. Combining a C18 phase with a hydrophilic end-capping, AquaSil™ C18 columns offer a unique material for reversed phase chromatography, offering alternative selectivity, up to twice the retention for polar compounds, and no phase collapse under 100% aqueous conditions. Ideal for use with LC/MS, these columns maintain selectivity with reduced concentrations of buffers and additives.
  • Compatible with 100% aqueous mobile phase
  • Good results with low buffer concentrations
  • Stable for LC/MS applications