U heeft gezocht op: Aardingskabels
11  results were found
SearchResultCount:"11"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (115-3046)
Leverancier: DESCO
Omschrijving: This provides a storage terminal for the wrist cord when the workstation is unoccupied.
UOM: 1 * 1 ST
Catalogus nummer: (115-3037)
Leverancier: DESCO
Omschrijving: The MagSnap® 360™ dual wire coil cord offers a 360° rotation and magnetic connection between the wristband and coil cord.
UOM: 1 * 1 ST
Leverancier: S.C.A.T.
Omschrijving: In order to prevent static discharge with fluids in plastic containers, the contents of the waste receptacles can be secured with ground connection. T...
Catalogus nummer: (115-3045)
Leverancier: DESCO
Omschrijving: The Desco Europe Slim Earth Bonding Point Bar (or Common Ground Point) has been designed to be installed underneath bench tops where it will be access...
UOM: 1 * 1 ST
Catalogus nummer: (115-3047)
Leverancier: DESCO
Omschrijving: Daisy-chains two worksurface mats for the monitoring of two workstations with the 724 workstation monitor.
UOM: 1 * 1 ST
Leverancier: Metro International
Omschrijving: These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conj...
Leverancier: Bohlender
Omschrijving: For earth connection of vessels and components made of static dissipative materials.
Catalogus nummer: (115-3044)
Leverancier: DESCO
Omschrijving: The EBP bar should be fixed to the edge of a bench or other suitable work equipment within reach and accessible to all connecting ground cords, but wh...
UOM: 1 * 1 ST
Leverancier: DESCO
Omschrijving: Coil cord with 2,5 mm polyurethane insulation, conductor is high strength 7-end duplex tinsel for strength and flexibility.
Catalogus nummer: (115-3069)
Leverancier: DESCO
Omschrijving: This cord with low profile helps keep employees from tripping on cord connection.
UOM: 1 * 1 ST
Catalogus nummer: (115-3056)
Leverancier: DESCO
Omschrijving: The Universal snap mounting kit consists of 10 mm stud and socket, threaded backing washer, conical nut, long and short screw, ESD common ground point...
UOM: 1 * 1 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 11 of 11
no targeter for Bottom