Overpack protection cover, PIG®
Leverancier: New Pig
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIG®
121-0157EA 154 EUR
121-0157
Overpack protection cover, PIG®
Deksels
Durable cover adds UV protection and visibility to PIG overpack spill kit.
  • Durable PVC-laminated polyester cover
  • Elasticised bottom to fit PIG® spill kits in 246- and 360-litre overpacks
  • Keeps water, snow, dirt and debris off overpack lid and resists weathering and UV
  • Features 230×280 mm see-through pocket for identification label
  • High visibility colour to make kit easy to locate