Barcode reader, PluraScan™, WHEATON®
Leverancier: Wheaton
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WHEAW986010EA 17530 EUR
WHEAW986010
Barcode reader, PluraScan™, WHEATON®
This sophisticated barcode reader for 2D data matrix is capable of reading bar codes from a wide range of manufacturer and label printers.
  • Integrated frost reduction system allows multiple racks to be read in a single session
  • Flexible software integration
It is ready to be used with the Wheaton KeepIT® boxes for use with Wheaton CryoELITE® vials and Cryule® Ampules, yet is versatile and can be adapted for use with multiple different rack and box formations.