Cut resistant gloves, HyFlex® 11-501
Supplier: Ansell
Certificates
About this item
These knitted Lycra®/Kevlar® liners with nitrile foam coating on the palm are cut and heat resistant, allowing versatile use.
- Ansell Intercept Technology™ improves mechanical performance and makes gloves resistant to heat and molten splashes
- Perfect fit, follow the hands' movements
- Silicone-free
- Anti-static
Ideal for workers who need high levels of protection with optimal productivity, e.g. in automotive (press and body shop, welding), metal fabrication, white goods and the glass industry.
Certifications: EN 388:2016 (3X41D), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- ANSE11-501/9
- ANSE11-501/8
- ANSE11-501/6
- ANSE11-501/10
- ANSE11-501/7
- Model:
- 11-501
- 11-501
- 11-501
- 11-501
- 11-501
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®/Lycra®
- Kevlar®/Lycra®
- Kevlar®/Lycra®
- Kevlar®/Lycra®
- Kevlar®/Lycra®
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3X41D), EN 407 (X1XXXX)
- EN 388:2016 (3X41D), EN 407 (X1XXXX)
- EN 388:2016 (3X41D), EN 407 (X1XXXX)
- EN 388:2016 (3X41D), EN 407 (X1XXXX)
- EN 388:2016 (3X41D), EN 407 (X1XXXX)
- Anti-static:
- +
- +
- +
- +
- +
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPkAvailabilityPrice
- Supplier Article Number: 11-501/6Local Article Number: ANSE11-501/6Specifications:
More
Specifications
Model11-501Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialKevlar®/Lycra®Gauge13Cuff styleKnit wristEN normEN 388:2016 (3X41D), EN 407 (X1XXXX)Anti-static+ - Supplier Article Number: 11-501/7Local Article Number: ANSE11-501/7Specifications:
More
Specifications
Model11-501Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialKevlar®/Lycra®Gauge13Cuff styleKnit wristEN normEN 388:2016 (3X41D), EN 407 (X1XXXX)Anti-static+ - Supplier Article Number: 11-501/8Local Article Number: ANSE11-501/8Specifications:
More
Specifications
Model11-501Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialKevlar®/Lycra®Gauge13Cuff styleKnit wristEN normEN 388:2016 (3X41D), EN 407 (X1XXXX)Anti-static+ - Supplier Article Number: 11-501/9Local Article Number: ANSE11-501/9Specifications:
More
Specifications
Model11-501Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialKevlar®/Lycra®Gauge13Cuff styleKnit wristEN normEN 388:2016 (3X41D), EN 407 (X1XXXX)Anti-static+ - Supplier Article Number: 11-501/10Local Article Number: ANSE11-501/10Specifications:
More
Specifications
Model11-501Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialKevlar®/Lycra®Gauge13Cuff styleKnit wristEN normEN 388:2016 (3X41D), EN 407 (X1XXXX)Anti-static+