Autoclaves, semi-automatic, M series
Leverancier: Tuttnauer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TUTTTU2340/M TUTTTU3870/M
Autoclaves, semi-automatic, M series
Sterilisatoren Autoclaven (stoomsterilisatoren)
These models feature double locking safety device with steam pressure activation prevents door from opening while chamber is pressurised.
  • Features insufficient water detector with automatic shutoff
  • Door failure protection prevents door from opening at high pressure
  • Automatic safety shutoff prevents overheating
  • Maintenance free with reduced running cost
Durable 316L type stainless steel chamber and electro-polish finish door.