Conductivity probes, sensION
Leverancier: Hach Lange
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH5197500EA 692 EUR
HACH5197500
Conductivity probes, sensION
Graphite, 4-pole conductivity probe with fixed cable and 5-pin circular connector compatible with Hach sensION meters.
  • Accurate
  • Easy to use, intuitive operation
  • Rugged and durable: Noryl sensor body withstand the rigors of outdoor testing and frequent use
  • Flexible: Suitable for both field and lab use
Leveringsinformatie: Includes 1000 µS/cm conductivity standard (100 ml), and user manual.