Wipes for disinfecting solutions, Bode X-Wipes
Leverancier: Bode
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BODE976710EA 0 EUR
BODL9805242 BODE976710 BODE980610 BODE980524 BODL980556_U BODE981373 BODE981372 BODE981371 BODL980584 BODE981370 BODE980583 115-2452 BODE976690_U
Wipes for disinfecting solutions, Bode X-Wipes
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Desinfectiedoekjes
These universally applicable fleece wipes can be used in combination with all liquid BODE surface disinfectants.
  • As the wipes are disposable, this reduces preparation time and ensures a high standard of hygiene
  • High quality, tear-resistant and absorbent PET fleece guarantees optimal disinfectant delivery
  • Reliable protection of solution from contaminants and evaporation
  • Sturdy dispenser with carrying handle
  • Coloured dispensing system for improved differentiation of applications or products
Ideal for disinfecting surfaces in all areas where a maximum level of hygiene, efficiency and convenience is required. The wipes are especially recommended for areas that must be cleaned several times per day. In the instrument area, they are ideal for preliminary cleaning and disinfection of medical devices that cannot be submerged (such as ultrasonic transducers and probes), as well as preliminary cleaning of flexible endoscopes.
BODE X-Wipes dispensing system: Each refillable dispensing system is composed of an X-Wipes dispenser filled with a dry roll of fleece wipes containing BODE X-Wipes that can be torn off individually, as well as a coloured dispensing system with a protective cap (integrated into the lid) and labels for identification (in German, English, French, Czech and Turkish).
Depending on the requirements, the system is available in two sizes, either as a dispenser or as a space saving canister. The dispenser can be equipped with rolls of 90 or 40 individual fleece wipes, and the canister can be equipped with rolls of 30 individual fleece wipes. Depending on the size, these are filled with 2,5 L (90-wipe roll), 1,5 L (40-wipe roll) or a minimum of 750 ml (30-wipe roll) of a surface disinfectant. A safety label used for identification is delivered with each roll.
Roll of fleece wipes in film bag: The dry roll of fleece wipes is also available in an innovative film bag. The solution being used with this item is poured directly into the plastic film bag with the roll of fleece wipes. This prevents any contact between the disinfectant-saturated fleece wipes and the inside of the dispenser.
The bag is made of sturdy, tear-resistant film and is supplied with a dispenser lid. Using the film bag requires very little effort: The film bag can simply be disposed of, and the wipe dispenser does not have to be reprocessed after use (with the exception of the cover lid). Prevents any contaminants being transferred from the wipe dispenser to the solution being used. Shelf life: Up to 4 weeks after filling (with a properly sealed protective lid).
Certificaten: Shelf life: Up to 4 weeks after filling (with a properly sealed protective lid)
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904