Reference electrode filling solutions
Leverancier: Mettler-Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
METT51343180EA 8.3 EUR
METT51343180 METT51343184 METT51343182 METT51350082 METT51350072 METT51350084 METT51350074 662-2886 METT51350076
Reference electrode filling solutions
Elektrolyten Vuloplossingen voor elektrodes
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.