Winter rain jacket, Adelans 191A
Leverancier: Sioen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIOE191A/N15/SEA 87.5 EUR
SIOE191A/N15/S SIOE191A/N15/1XL SIOE191A/N15/3XL SIOE191A/N15/2XL SIOE191A/N15/M SIOE191A/N15/L
Winter rain jacket, Adelans 191A
Werkkleding
Water- and windproof jacket made of 100% polyester fabric with PVC coating. With taped seams, fixed polyamide lining and detachable fleece lining. Provides good protection against the cold.
  • Straight collar with foldaway hood
  • Zip closure under flap with press studs
  • Two inset pockets with zip closure
  • Raglan sleeves, sleeve narrowing by touch and close fastening
  • Elasticated drawstring in waist and hem
  • Fixed polyamide lining with inside pocket
  • Knitted wind cuffs in sleeves
Back length: 85 cm
Weight: 270 g/m²
Certificaten: Detachable lining only: EN 14058 (1XXXX)