Winter rain jacket, Dover 4893
Supplier: SIOEN
Certificates
About this item
Water- and windproof jacket made of Flexothane® Essential: 100% polyester knitting with PU coating. With detachable fleece lining in body and hood and quilted sleeves.
- Fixed hood, zip closure under flap with press studs
- Two patched pockets with flap closed by press studs
- Raglan sleeves
- High frequency welded seams
- Knitted wind cuffs at end of sleeves
- Detachable lining can not be worn separately
Weight: 170 g/m²
Certifications: EN 343 (3-1)
Specifications
- Catalog No:
- SIOE4893/B90/1XL
- SIOE4893/A41/S
- SIOE4893/A41/M
- SIOE4893/A41/L
- SIOE4893/A41/3XL
- SIOE4893/A41/2XL
- SIOE4893/B90/S
- SIOE4893/A41/1XL
- SIOE4893/B90/M
- SIOE4893/Y58/M
- SIOE4893/B90/L
- SIOE4893/Y58/S
- SIOE4893/B90/3XL
- SIOE4893/B90/2XL
- SIOE4893/Y58/L
- SIOE4893/Y58/2XL
- SIOE4893/Y58/3XL
- SIOE4893/Y58/1XL
- Model:
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- 4893 Dover
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- Weight:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
Specifications
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: 4893/A41/SLocal Article Number: SIOE4893/A41/SSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/A41/MLocal Article Number: SIOE4893/A41/MSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/A41/LLocal Article Number: SIOE4893/A41/LSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/A41/1XLLocal Article Number: SIOE4893/A41/1XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/A41/2XLLocal Article Number: SIOE4893/A41/2XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/A41/3XLLocal Article Number: SIOE4893/A41/3XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/SLocal Article Number: SIOE4893/B90/SSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/MLocal Article Number: SIOE4893/B90/MSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/LLocal Article Number: SIOE4893/B90/LSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/1XLLocal Article Number: SIOE4893/B90/1XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/2XLLocal Article Number: SIOE4893/B90/2XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/B90/3XLLocal Article Number: SIOE4893/B90/3XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/SLocal Article Number: SIOE4893/Y58/SSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/MLocal Article Number: SIOE4893/Y58/MSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/LLocal Article Number: SIOE4893/Y58/LSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/1XLLocal Article Number: SIOE4893/Y58/1XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/2XLLocal Article Number: SIOE4893/Y58/2XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m² - Supplier Article Number: 4893/Y58/3XLLocal Article Number: SIOE4893/Y58/3XLSpecifications:
More
Specifications
Model4893 DoverGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaRain (waterproof), ColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyester knitting with PU coatingWeight170 g/m²