Netherlands
Order Entry
Water baths, unstirred, SUB Aqua Pro series
undefined
Water baths, unstirred, SUB Aqua Pro series
  •  
    Type
    Temperature Range
    Temp. stability
    Heating capacity
    Capacity
    Bath W×D×H
  • 462-0474
    SAP2
    SAP2
    RT* +5...99 °C
    ± 0,2 °C
    0,25 kW
    2 L
    131×117×132 mm
    1 * 1 ST
    1 * 1 ST - 1 170,00 €
    1 170,00 €
    1 * 1 ST
       1 170,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0475
    SAP2S
    SAP2S (shallow)
    RT* +5...99 °C
    ± 0,2 °C
    0,35 kW
    2 L
    289×139×47 mm
    1 * 1 ST
    1 * 1 ST - 1 180,00 €
    1 180,00 €
    1 * 1 ST
       1 180,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0476
    SAP5
    SAP5
    RT* +5...99 °C
    ± 0,2 °C
    0,35 kW
    5 L
    289×131×132 mm
    1 * 1 ST
    1 * 1 ST - 1 230,00 €
    1 230,00 €
    1 * 1 ST
       1 230,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0477
    SAP12
    SAP12
    RT* +5...99 °C
    ± 0,2 °C
    0,8 kW
    12 L
    306×281×132 mm
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0478
    SAP18
    SAP18
    RT* +5...99 °C
    ± 0,2 °C
    1,4 kW
    18 L
    281×485×132 mm
    1 * 1 ST
    1 * 1 ST - 1 520,00 €
    1 520,00 €
    1 * 1 ST
       1 520,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0479
    SAP26
    SAP26
    RT* +5...99 °C
    ± 0,2 °C
    1,4 kW
    26 L
    278×481×182 mm
    1 * 1 ST
    1 * 1 ST - 1 590,00 €
    1 590,00 €
    1 * 1 ST
       1 590,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0480
    SAP34
    SAP34
    RT* +5...99 °C
    ± 0,2 °C
    1,8 kW
    34 L
    281×635×180 mm
    1 * 1 ST
    1 * 1 ST - 2 500,00 €
    2 500,00 €
    1 * 1 ST
       2 500,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • 462-0481
    SAPD
    SAPD
    RT* +5...99 °C
    ± 0,2 °C
    1,15 kW
    5; 12 L
    281×131×150
    306×281×150 mm
    1 * 1 ST
    1 * 1 ST - 2 260,00 €
    2 260,00 €
    1 * 1 ST
       2 260,00 €
    Accessories
    Specifications
    Environmentally Preferable
Stainless steel test tube racks
  •  
    Seq
    Description
    For
    Pk
  • 005
    Stainless steel test tube rack for 16 mm Ø tubes
    36 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 003
    Stainless steel test tube rack for 10 mm Ø tubes
    84 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 006
    Stainless steel test tube rack for 19 mm Ø tubes
    32 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 007
    Stainless steel test tube rack for 25 mm Ø tubes
    18 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 008
    Stainless steel test tube rack for 30 mm Ø tubes
    12 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 001
    Stainless steel rack for 0.5 ml microtubes
    105 tubes
    1
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
  •  
    Seq
    Description
    For
    Pk
  • 002
    Stainless steel rack for 1.5 ml microtubes
    65 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
  •  
    Seq
    Description
    For
    Pk
  • 004
    Stainless steel test tube rack for 13 mm Ø tubes
    55 tubes
    1
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
Stainless steel raised shelves
  •  
    Seq
    Description
    For
    Pk
  • 050
    Raised half shelf with perforated mesh
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 205,00 €
    205,00 €
  •  
    Seq
    Description
    For
    Pk
  • 051
    Raised half shelf with perforated mesh
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
  •  
    Seq
    Description
    For
    Pk
  • 052
    Raised half shelf with perforated mesh
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 286,00 €
    286,00 €
  •  
    Seq
    Description
    For
    Pk
  • 053
    Raised half shelf with perforated mesh
    SAP34, (JB Aqua 34 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
Stainless steel base trays
  •  
    Seq
    Description
    For
    Pk
  • 062
    Stainless steel base tray**
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    1
    1 * 1 ST
    1 * 1 ST - 130,00 €
    130,00 €
  •  
    Seq
    Description
    For
    Pk
  • 063
    Stainless steel base tray**
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    1
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
  •  
    Seq
    Description
    Pk
    Price
  • 060
    Stainless steel base tray**, for SAP2
    1
    44,00
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
  •  
    Seq
    Description
    Pk
  • 061
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    1
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
Replacement lids and base trays
  •  
    Seq
    Description
    For
    Pk
  • 041
    Gabled stainless steel lid
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    1
    1 * 1 ST
    1 * 1 ST - 294,00 €
    294,00 €
  •  
    Seq
    Description
    For
    Pk
  • 043
    Gabled stainless steel lid
    SAP34, SUB36
    1
    1 * 1 ST
    1 * 1 ST - 381,00 €
    381,00 €
  •  
    Seq
    Description
    For
    Pk
  • 021
    Replacement base trays, polycarbonate
    SAP2S, SAP5, SAPD, JBA5 , JBN5
    1
    1 * 1 ST
    1 * 1 ST - 66,90 €
    66,90 €
  •  
    Seq
    Description
    For
    Pk
  • 025
    Replacement lids, polycarbonate
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    1
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
  •  
    Seq
    Description
    For
    Pk
  • 022
    Replacement base trays, polycarbonate
    JBA12, JBN12, SAP12 and SAPD
    1
    1 * 1 ST
    1 * 1 ST - 70,40 €
    70,40 €
  •  
    Seq
    Description
    For
    Pk
  • 040
    Gabled stainless steel lid
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 267,00 €
    267,00 €
  •  
    Seq
    Description
    For
    Pk
  • 023
    Replacement base trays, polycarbonate
    SAP18, SAP26, JBA18, JBN18 , JBN26
    1
    1 * 1 ST
    1 * 1 ST - 73,90 €
    73,90 €
  •  
    Seq
    Description
    For
    Pk
  • 042
    Gabled stainless steel lid
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    1
    1 * 1 ST
    1 * 1 ST - 330,00 €
    330,00 €
  •  
    Seq
    Description
    For
    Pk
  • 031
    Flat lid with 4 ring sets
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 492,00 €
    492,00 €
  •  
    Seq
    Description
    For
    Pk
  • 030
    Flat lid with 2 ring sets
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 320,00 €
    320,00 €
  •  
    Seq
    Description
    For
    Pk
  • 032
    Flat lid with 6 ring sets
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    1
    1 * 1 ST
    1 * 1 ST - 682,00 €
    682,00 €
  •  
    Seq
    Description
    For
    Pk
  • 033
    Flat lid with 8 ring sets
    SAP34, SUB36
    1
    1 * 1 ST
    1 * 1 ST - 764,00 €
    764,00 €
  •  
    Seq
    Description
    For
    Pk
  • 024
    Replacement lids, polycarbonate
    SAP2
    1
    1 * 1 ST
  •  
    Seq
    Description
    For
    Pk
  • 020
    Replacement base trays, polycarbonate
    SAP2
    1
    1 * 1 ST
    1 * 1 ST - 56,30 €
    56,30 €