Ladies shorts, C.I., 294 PR54, Design C, navy blue
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KWIN846539231C58EA 0 EUR
KWIN846539231C58 KWIN846539440C60 KWIN846539331C60 KWIN846539520C60 KWIN846539520C62 KWIN846539231C50 KWIN846539231C52 KWIN846539231C54 KWIN846539231C56 KWIN846539131C44 KWIN846539440C58 KWIN846539231C60 KWIN846539331C56 KWIN846539520C54 KWIN846539440C56 KWIN846539131C48 KWIN846539331C58 KWIN846539131C46 KWIN846539440C54 KWIN846539520C56 KWIN846539331C52 KWIN846539520C58 KWIN846539440C52 KWIN846539331C54 KWIN846539440C50 KWIN846539331C50 KWIN846539231C62 KWIN846539210C62 KWIN846539730C50 KWIN846539730C52 KWIN846539131C52 KWIN846539730C44 KWIN846539131C50 KWIN846539730C46 KWIN846539131C56 KWIN846539331C48 KWIN846539730C48 KWIN846539131C54 KWIN846539440C48 KWIN846539440C46 KWIN846539331C44 KWIN846539131C58 KWIN846539440C44 KWIN846539331C46 KWIN846539941C46 KWIN846539941C48 KWIN846539131C62 KWIN846539941C50 KWIN846539131C60 KWIN846539941C52 KWIN846539941C54 KWIN846539539C62 KWIN846539539C60 KWIN846539231C48 KWIN846539520C50 KWIN846539520C52 KWIN846539941C56 KWIN846539941C58 KWIN846539231C44 KWIN846539231C46 KWIN846539539C56 KWIN846539941C60 KWIN846539941C62 KWIN846539539C54 KWIN846539539C58 KWIN846539520C44 KWIN846539520C46 KWIN846539539C52 KWIN846539520C48 KWIN846539440C62 KWIN846539539C50 KWIN846539940C58 KWIN846539940C56 KWIN846539539C44 KWIN846539539C48 KWIN846539539C46 KWIN846539940C62 KWIN846539940C60 KWIN846539530C58 KWIN846539530C56 KWIN846539530C54 KWIN846539530C52 KWIN846539941C44 KWIN846539530C62 KWIN846539530C60 KWIN846539530C48 KWIN846539530C46 KWIN846539530C44 KWIN846539210C54 KWIN846539640C52 KWIN846539210C52 KWIN846539730C60 KWIN846539640C54 KWIN846539210C58 KWIN846539640C56 KWIN846539730C62 KWIN846539210C56 KWIN846539640C58 KWIN846539730C54 KWIN846539730C56 KWIN846539640C60 KWIN846539730C58 KWIN846539210C60 KWIN846539640C62 KWIN846539530C50 KWIN846539940C48 KWIN846539210C48 KWIN846539940C46 KWIN846539940C44 KWIN1008469405840 KWIN846539640C44 KWIN846539640C46 KWIN846539210C46 KWIN846539640C48 KWIN846539210C44 KWIN1008469405838 KWIN846539210C50 KWIN846539640C50 KWIN846539940C54 KWIN846539940C52 KWIN846539940C50 KWIN846539331C62
Ladies shorts, C.I., 294 PR54, Design C, navy blue
Werkkleding
Short trousers made of 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring
Weight: 260 g/m²
These trousers can be customised. You can chose between 12 profile colours and 13 piping colours.