Long johns, Devold® 767 U
Leverancier: Fristads Kansas Group
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Devold®
KWIN100614940MEA 0 EUR
KWIN100614940M KWIN100614940L KWIN1006149402XL KWIN100614940XL KWIN100614940S
Long johns, Devold® 767 U
Werkkleding
Functional long johns made of 100% wool. Provide good protection against cold, radiant heat, flame and metal splash. The garment has a smooth outer layer offering the best protection, and a brushed inside layer to maximise the insulating properties.
  • Shrink-treated
  • Zirpro treated to give extra fire retardancy
  • Wool is voluminous and airy and has excellent insulation properties
  • Wool can absorb significant amounts of moisture without feeling wet
  • With fly
  • Elasticated waist and leg ends
Weight: 470 g/m²
Certificaten: EN ISO 11612 (A2 B2 C2) and EN 61482-1-2 (class 1)