VWR® DNA Molecular Weight Marker, High-Range
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
E255-150UGEA 1080 EUR
E255-150UG
VWR® DNA Molecular Weight Marker, High-Range
Reagentia voor elektroforese Molecuulgewichtmerkers
High-Range DNA MW (molecular weight) marker offers 10 fragments ranging from 1503 to 48 502 bp. There is sufficient material for 75 to 150 assays.
  • Optimal for evaluation fragments >10 kb
  • Easy band identification