Emergency shower for installation over doors
Leverancier: Broen Lab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
118-1947EA 1160 EUR
BROE17.087.509
Emergency shower for installation over doors
Powder coated safety shower unit made from brass and steel, for installation over doors.
  • Ball valve with quick-assembly system on wall flange
  • With pull rod
  • Jet emergency shower head made from PPO, with improved spray pattern, self-draining
  • Operating controls can be fitted on the right or left
  • Easy to mount and install
  • Pre-mounted valve
Weight: ~4,5 kg
Connection: G ¾" (side)
Min. working pressure: 1 bar
Certificaten: EN 15154-1; ANSI Z358.1. TÜV certified