HPLC columns, Hypersil™ ODS C18
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Hypersil™
555-6150EA 755 EUR
555-6150 555-6151 555-6235 555-6152 555-6153 555-6154 555-6162 555-6127 555-6129 555-6167 555-6169 555-6163 555-6170 555-6171 555-6172 555-6173 554-5316 554-5315 554-5314 554-5313 555-0237 555-6138 555-6215 555-6216 555-0238 555-6217 555-6218 555-6134 555-6211 555-6179 555-6135 555-6136 555-6213 555-6174 555-6131 555-6132 555-6177 555-6210 555-6181 555-6183 555-6140 555-6180 555-6226 555-6149 555-6222 555-6146 555-6223 555-6224 555-6225 555-6186 555-6187 555-6143 555-6220 555-6144
HPLC columns, Hypersil™ ODS C18
Kolommen voor chromatografie
These ODS columns are suitable for non-polar and moderately polar analytes including acids, neutrals and lipophilic compounds. All column phases are endcapped.
  • C18 stationary phase for a broad range of applications