VWR® Q Path® Freeze Gel (Glue) Mounting Medium for Cryotomy
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® Q path®
07111245.EA 190 EUR
07111245.
VWR® Q Path® Freeze Gel (Glue) Mounting Medium for Cryotomy
Kleurstoffen Microscopy and staining reagents
Hoogviskeuze montage media voor Cryotomie. Minder viskeus dan O.C.T.