O-Phenylenediamine (OPD tablets), ultrapure
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J348-50TEA 131 EUR
VWRCJ348-50T
O-Phenylenediamine (OPD tablets), ultrapure
OPD tablets are widely used to detect horseradish peroxidase activity in ELISA and solution assays. Absorbs at 492 nm. Each tablet contains 15 mg of OPD.