VWR® Q Path® Isomount 2000, Mounting Media
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® Q path®
05547536.EA 50 EUR
05547536. 05547535.
VWR® Q Path® Isomount 2000, Mounting Media
Kleurstoffen Microscopy and staining reagents
Mounting media based on isopropyl alcohol. Compatible with most solvent replacing agents. Suitable for manual and automatic mounting of slides. PET bottle of 500 ml.
  • Less toxic mounting
  • Approved quality
  • Drying time: 20 m