Vacuum manifold and accessories
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
The multi-well plate vacuum manifold is an anodised aluminium manifold that has been designed and optimised for the vacuum filtration of AcroPrep™, AcroPrep Advance, and AcroWell™ multi-well filter plates.
- Supplied complete with the necessary O-ring and gasket; control block includes vacuum pressure gauge, vacuum metering valve, vacuum release valve, and ¼" hose barb for vacuum line attachment
- Vacuum manifold unit includes a Delrin® plastic spacer block designed to accommodate standard 350 μl receiver plates and optimised to reduce space between the receiver plate and the filter plate during vacuum filtration
- Optional spacer block available for use with receiver plates
- Adapter collar holds filter plates tightly to receiver plates for centrifugation
Related Information
Pall Laboratory AcroPrep Filter-Plates Pall Laboratory Lab essentials optimizing sample preparation and product purification
Product Literature
Pall Lysate Clearance
Application Notes
PALL5017_TDS_EN
Datasheet
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Multi-well filter plate vacuum manifoldSupplier Article Number: 5017Local Article Number: PALL5017Specifications:
Accessories
Specifications
Environmentally Preferable - Receiver plate spacer block, 1 mlSupplier Article Number: 5014-NLocal Article Number: PALL5014-NSpecifications:
Accessories
Specifications
Environmentally Preferable - Receiver plate spacer block, 350 µlSupplier Article Number: 5015-NLocal Article Number: PALL5015-NSpecifications:
Accessories
Specifications
Environmentally Preferable