Drum tap, zinc
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA2796EA 44.3 EUR
MSGA2796
Drum tap, zinc
Kranen en gootstenen Kranen
This fine dosing tap is made of zinc injection moulding.
  • Self closing
  • Flame guard
  • PTFE seal