5-Bromo-o-toluic acid ≥97%
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H60516.06EA
117
EUR
H60516.06
5-Bromo-o-toluic acid ≥97%
5-Bromo-o-toluic acid
Formule:
C₈H₇BrO₂ MW: 215,05 g/mol Smeltpunt: 167…171 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00267350 CAS nummer: 79669-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...