Surgical masks, PFR P2, PE/PP/PU
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ANSM62408EA 89.8 EUR
ANSM62408
Surgical masks, PFR P2, PE/PP/PU
Latex surgical mask with Polypropylene filter to ensure the best filtration.
  • Maintaining tight against the face mask by two polyurethane ties
  • Ultrasonic welding Sontec II
  • Inner layer polypropylene+polyethylene for high comfort
  • Without fiberglass, odourless
NaCl Penetration: 0.284 %
Paraffin penetration : 2,28%
Facial leakage: 0.47%
Delta P of less than 2 mm H₂O
Skin tolerance test and Nelson test
Certificaten: EN 149