GL-reductions, for tubing, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLD784-01EA 37.7 EUR
BOHLD784-01 BOHLD784-24 BOHLD784-03 BOHLD784-04 BOHLD784-16 BOHLD784-06 BOHLD784-08
GL-reductions, for tubing, BOLA
Fittingen en connectoren
Connector, GL screw thread reduction, with black screw cap made of PPS with PTFE/FPM sealing, with movable reduction body made of PTFE for transition to GL threads.
  • Applicable for connecting or inserting tubing, tubes or probes
  • Temperature resisitance from –200 to +250 ºC
  • Excellent chemical resistance
  • FDA conformity
The body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable. The product is only exposed to PTFE.