Pasteur pipettes, graduated, WHEATON®
Leverancier: Wheaton
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LSUK726128EA 51.8 EUR
LSUK726128 LSUK711117S/20 LSUK726129 LSUK711118S/20
Pasteur pipettes, graduated, WHEATON®
Pipetten Pasteur-/transferpipetten
LDPE, transparent, sterile.
  • Pipettes with bulb and graduations
  • Used for safe and easy transfer of liquids and samples
  • Narrow stem pipette suitable for use as freeze vial when bulb heat sealed