About this item
Unistats® are the ideal solution for fast and precise thermal control of externally connected applications. These compact units enable previously unachievable performance, offer rapid temperature change and allow use over a wide temperature range without requiring a fluid change. There are over 60 models to choose from in sleek tower housings, or flat-build with cooling powers from 0,7 to 130 kW. Whatever the application, Unistats® provide professional scale up offering the same stable process conditions from the development lab to production systems. Units have a range of safety functions, provide flexible communication via USB and LAN interfaces and give comprehensive warning alarms. Units are ideal for use with reactor systems, autoclaves, pilot and mini plant systems, scale up developments, double wall reactors, distillation systems, material testing and use in combinational chemistry and the semiconductor industry.
- Working temperatures from –120 to +425 °C
- Multilingual, interactive Pilot ONE® controller with Plug & Play technology, 14,5 cm touch screen and easy menu navigation and graphic capabilities
- Highly accurate, intelligent temperature control
- Maximum process stability and reproducibility
- The fastest heating and cooling rates
- Increased thermal fluid life
Units comply with DIN 12876; rated Safety Class III
Air cooled models
Product Information
- Catalog #Supplier #TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityCapacityAvailabilityPrice
- 461-10511030.0007.01Petite Fleur-40...+200 °C±0,01 °C1,5 kW0,48; 0,48; 0,45; 0,27; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1030.0007.01Local Article Number: HUBE1030.0007.01Specifications:
Accessories
Specifications
Environmentally Preferable - HUBE1002.0045.011002.0045.01Unistat 405-45...+250 °C±0,01 °C3,0 kW1; 1; 1; 0,6; 0,15 kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1002.0045.01Local Article Number: HUBE1002.0045.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-08321030.0004.01Petite Fleur-eo-40...+200 °C±0,01 °C1,5 kW0,48; 0,48; 0,45; 0,27; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1030.0004.01Local Article Number: HUBE1030.0004.01Specifications:
Accessories
Specifications
Environmentally Preferable - HUBE1000.0037.011000.0037.01Unistat Tango-45...+250 °C±0,01 °C3,0 kW0,7; 0,7; 0,7; 0,4; 0,06 kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1000.0037.01Local Article Number: HUBE1000.0037.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-07431030.0001.01Petite Fleur-NR-40...+200 °C±0,01 °C1,5 kW0,48; 0,48; 0,45; 0,27; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1030.0001.01Local Article Number: HUBE1030.0001.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-10531041.0004.01Grande Fleur-eo-40...+200 °C±0,01 °C1,5 kW0,6; 0,6; 0,6; 0,35; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1041.0004.01Local Article Number: HUBE1041.0004.01Specifications:
Accessories
Specifications
Environmentally Preferable
Water cooled models
Product Information
- Catalog #Supplier #TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityCapacityAvailabilityPrice
- Supplier Article Number: 1032.0011.01Local Article Number: HUBE1032.0011.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-05621030.0003.01Petite Fleur w-NR-40...+200 °C±0,01 °C1,5 kW0,48; 0,48; 0,45; 0,27; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1030.0003.01Local Article Number: HUBE1030.0003.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-10541041.0007.01Grande Fleur w-40...+200 °C±0,01 °C1,5 kW0,6; 0,6; 0,6; 0,35; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1041.0007.01Local Article Number: HUBE1041.0007.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-10551041.0010.01Grande Fleur w-eo-40...+200 °C±0,01 °C1,5 kW0,6; 0,6; 0,6; 0,35; 0,04; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1041.0010.01Local Article Number: HUBE1041.0010.01Specifications:
Accessories
Specifications
Environmentally Preferable - HUBE1002.0046.011002.0046.01Unistat 405w-45...+250 °C±0,01 °C3,0 kW1,3; 1,3; 1,3; 0,7; 0,15; -; - kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1002.0046.01Local Article Number: HUBE1002.0046.01Specifications:
Accessories
Specifications
Environmentally Preferable - HUBE1000.0039.011000.0039.01Unistat Tango w-45...+250 °C±0,01 °C3,0 kW0,7; 0,7; 0,7; 0,4; 0,06 kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1000.0039.01Local Article Number: HUBE1000.0039.01Specifications:
Accessories
Specifications
Environmentally Preferable - HUBE1000.0040.011000.0040.01Unistat Tango wl-45...+250 °C±0,01 °C3,0 kW0,7; 0,7; 0,7; 0,4; 0,06 kW1,5 lLog in for availability1 * 1 STSupplier Article Number: 1000.0040.01Local Article Number: HUBE1000.0040.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-60141021.0016.01Unistat 430w-FB-40...+250 °C±0,01 °C4 kW3,5; 3,5; 3,5; 2,2; 0,3; - kW4,1 lLog in for availability1 * 1 STSupplier Article Number: 1021.0016.01Local Article Number: HUBE1021.0016.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-60131021.0015.01Unistat 425w-FB-40...+250 °C±0,01 °C2 kW2,8; 2,8; 2,5; 1,9; 0,2; - kW5,0 lLog in for availability1 * 1 STSupplier Article Number: 1021.0015.01Local Article Number: HUBE1021.0015.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-60161023.0011.01Unistat 815w-FB-85...+250 °C±0,01 °C21,5; 1,5; 1,5; 1,5; 1,4; 0,2 kW4,5 lLog in for availability1 * 1 STSupplier Article Number: 1023.0011.01Local Article Number: HUBE1023.0011.01Specifications:
Accessories
Specifications
Environmentally Preferable - 461-60151021.0017.01Unistat 510w-FB-50...+250 °C±0,01 °C6 kW5,0; 5,0; 5,0; 2,8; 0,9; - kW5,0 lLog in for availability1 * 1 STSupplier Article Number: 1021.0017.01Local Article Number: HUBE1021.0017.01Specifications:
Accessories
Specifications
Environmentally Preferable