Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Transport swabs, NRSII™ Transwab® range
Leverancier: MEDICAL WIRE & EQUIPMENT
Transwab®
Design with swab preimmersed, ready for use with a self standing tube and a high visibility blue cap and swab, which is easy to find and retrive if accidentally dropped. Conforms to ISO 18593 and other national and international standards. Neutralisers allow accurate recovery of residual bacteria on food contact surfaces. Precise fill for accurate assessment of contamination levels.
  • Blue cap and swab which can be easily retrived if dropped
  • Reduced packaging and storage space
  • Ready to use
  • Ambient storage and transport
Order
Expand All / Collapse All
Omschrijving Kleur dop VPE VWR catalogus nummer Hoeveelheid
Swabs with 5 ml NRS blauw 50 MEDWMW784
Swabs with 10 ml buffered peptone water blauw 50 MEDWMW776
Swabs with 10 ml NRS blauw 50 MEDWMW785
Swabs with 10 ml Letheen broth blauw 50 MEDWMW792
Swabs with 10 ml maximum recovery diluent blauw 50 MEDWMW796
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid
Een nieuwe uitstraling en een hernieuwde belofte
Gebouwd op de kenmerken & middelen die u kent, geoptimaliseerd met de innovatie van Avantor ®. Ons nieuwe merk symboliseert flexibiliteit en ons vermogen om ons aan de behoeften van de klant aan te passen.
KOM MEER TE WETEN BEGIN MET WINKELEN