Eye and face shower, for wall-mounting
Leverancier: Broen Lab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BROE17.928.009EA 1300 EUR
BROE17.928.009
Eye and face shower, for wall-mounting
Eye/face shower with two heads in brass with chemical resistant polycoating and stainless steel bowl.
  • With PUSH handle
  • Wall-mounted for exposed pipe work
  • Water amount is regulated by a built-in FLOWFIX (26 l/min)
Weight: ~4,5 kg
Connection: G1/2" female inlet
Min. working pressure: 1,5 bar
Certificaten: EN 15154-2; ANSI Z358.1. TÜV certified