Netherlands
Order Entry
Accessories for oil diffusion pump systems, DP, ILMVAC
Accessories for oil diffusion pump systems, DP, ILMVAC
undefined
Accessories for oil diffusion pump systems, DP, ILMVAC
  •  
    Pk
  • Multi-range vacuum gauge VMpro 1, DN16KF, 100 - 240 V 50/60 Hz
    Multi-range vacuum gauge VMpro 1, DN16KF, 100 - 240 V 50/60 Hz
    ILMV600092
    600092
    1
    1 * 1 ST
    1 * 1 ST - 857,00 €
    857,00 €
    1 * 1 ST
       857,00 €
     : 600092
     : ILMV600092
    Specifications
    Environmentally Preferable
  • Multi-range vacuum gauge with external display VMpro 3, chemical resistant, DN16KF, 100 - 240 V 50/60 Hz
    Multi-range vacuum gauge with external display VMpro 3, chemical resistant, DN16KF, 100 - 240 V 50/60 Hz
    ILMV600095
    600095
    1
    1 * 1 ST
    1 * 1 ST
     : 600095
     : ILMV600095
    Specifications
    Environmentally Preferable
  • Silicone oil, 0,5 l, XT 704
    Silicone oil, 0,5 l, XT 704
    ILMV800106
    800106
    1 L
    1 * 1 L
    1 * 1 L - 608,50 €
    608,50 €
    1 * 1 L
       608,50 €
     : 800106
     : ILMV800106
    Specifications
    Environmentally Preferable