Transport containers
Leverancier: Deltalab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
631-9780EA 12.7 EUR
631-9780 DELT900025 DELT979930
Transport containers
Verzenddoosjes voor objectglaasjes
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.