Cut resistant gloves, K-TEX® 930
Supplier: KCL by Honeywell
Certificates
About this item
These para-aramid gloves with natural latex coating have excellent flexibility, grip and resistance to cuts. Thanks to the design of the coating, even the finger tips are protected.
- Good grip, especially with moist parts
- Very good flexibility and high wearing comfort
- Good puncture resistance regarding metal (not syringes) and glass
- Allows working without user fatigue
Ideal for use in the glass and beverage industry, e.g. when sorting out broken glass in breweries or for handling of wet glass parts. Also suitable for use in laboratories (handling of non-contaminated Erlenmeyer flasks, putting together glass lines, disposal of broken glass) and in the waste disposal industry.
Colour: Yellow/blue
EN 388 (3444); Cat II
Certifications: EN 388 (3444); Cat. II
Specifications
- Catalog No:
- 112-8300
- KCHL930.8
- 112-2992
- KCHL930.7
- Model:
- 930
- 930
- 930
- 930
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Para-aramid
- Para-aramid
- Para-aramid
- Para-aramid
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3444)
- EN 388 (3444)
- EN 388 (3444)
- EN 388 (3444)
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPkAvailabilityPrice
- Supplier Article Number: 930.7Local Article Number: KCHL930.7Specifications:
More
Specifications
Model930Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialPara-aramidFinishPalm and fingertips coatedCuff styleKnit wristEN normEN 388 (3444) - Supplier Article Number: 930.8Local Article Number: KCHL930.8Specifications:
More
Specifications
Model930Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialPara-aramidFinishPalm and fingertips coatedCuff styleKnit wristEN normEN 388 (3444) - Supplier Article Number: 930.9Local Article Number: KCHL930.9Specifications:
More
Specifications
Model930Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialPara-aramidFinishPalm and fingertips coatedCuff styleKnit wristEN normEN 388 (3444) - Supplier Article Number: 930.10Local Article Number: KCHL930.10Specifications:
More
Specifications
Model930Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialPara-aramidFinishPalm and fingertips coatedCuff styleKnit wristEN normEN 388 (3444)