Cell culture media, Minimum Essential Medium (MEM)
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Biowest
L0476-500EA 12 EUR
L0476-500 P0450-N1L L0444-500 L0416-500 P0450-N10L P0451-N50L P0440-N10L P0515-N50L P0451-N5L P0440-N50L L0470-500 L0445-500 P0451-N1L P0515-N10L P0515-N1L L0415-500 P0451-N10L P0450-N50L L0440-500 P0440-N1L L0430-500 P0440-N5L L0475-500
Cell culture media, Minimum Essential Medium (MEM)
Celcultuurmedia
Minimum Essential Medium (MEM) with Earle's Balanced Salts is a modification of Eagle's earlier medium, Basal Medium Eagle (BME), containing higher concentrations of the essential nutrients. These media have demonstrated the ability to support a variety of normal and transformed cells in culture and contain Earle's Balanced Salts, which make them suitable for use in atmospheres charged with CO₂ gas.