Netherlands
Order Entry
Blades for Martor cutters
Blades for Martor cutters
Supplier:  MARTOR ARGENTAX
Certificates
undefined
Blades for Martor cutters
Supplier:  MARTOR ARGENTAX
Product Information
  • Catalog #
    Supplier #
    Material
    Thickness
    L×W
    Pk
    Availability
    Price
  • Trapezoidal, deep-edged, 2-sided ground-edge, rounded blade tips
    Trapezoidal, deep-edged, 2-sided ground-edge, rounded blade tips
    233-0192
    65232.70
    Steel
    0,63 mm
    50×19 mm
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 14,20 €
    14,20 €
    1 * 10 ST
       14,20 €
    Supplier Article Number: 65232.70
    Local Article Number: MARO65232.70
    Specifications:
    Specifications
    Environmentally Preferable
  • Hook blades, 2-sided ground-edge
    Hook blades, 2-sided ground-edge
    MARO56.70
    56.70
    Steel
    0,63 mm
    51×19 mm
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 21,50 €
    21,50 €
    1 * 10 ST
       21,50 €
    Supplier Article Number: 56.70
    Local Article Number: MARO56.70
    Specifications:
    Specifications
    Environmentally Preferable
  • Trapezoidal, deep-edged, 2-sided ground edge
    Trapezoidal, deep-edged, 2-sided ground edge
    233-2232
    5232.70
    Steel
    0,63 mm
    53×19 mm
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 13,00 €
    13,00 €
    1 * 10 ST
       13,00 €
    Supplier Article Number: 5232.70
    Local Article Number: MARO5232.70
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together