Netherlands
Order Entry
Cut resistant gloves, Showa 540D
Cut resistant gloves, Showa 540D
Supplier:  Showa
Certificates
undefined
Cut resistant gloves, Showa 540D
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5858
  • 112-5857
  • 112-5856
  • 112-5860
  • 112-5859
  • Model:
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • Finish:
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    Glove size
    Colour
    Length
    Packed
    Pk
    Availability
    Price
  • 112-5858
    540D-S
    S
    White
    210 - 260 mm
    10 pairs/pack
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 162,00 €
    162,00 €
    1 * 10 PAAR
       162,00 €
    Specifications:
    More
    Specifications
    Model
    Palm Fit 540D
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4241B)
  • 112-5857
    540D-M
    M
    White
    210 - 260 mm
    10 pairs/pack
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 162,00 €
    162,00 €
    1 * 10 PAAR
       162,00 €
    Specifications:
    More
    Specifications
    Model
    Palm Fit 540D
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4241B)
  • 112-5856
    540D-L
    L
    White
    210 - 260 mm
    10 pairs/pack
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 162,00 €
    162,00 €
    1 * 10 PAAR
       162,00 €
    Specifications:
    More
    Specifications
    Model
    Palm Fit 540D
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4241B)
  • 112-5859
    540D-XL
    XL
    White
    210 - 260 mm
    10 pairs/pack
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 162,00 €
    162,00 €
    1 * 10 PAAR
       162,00 €
    Specifications:
    More
    Specifications
    Model
    Palm Fit 540D
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4241B)
  • 112-5860
    540D-XXL
    XXL
    White
    210 - 260 mm
    10 pairs/pack
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 162,00 €
    162,00 €
    1 * 10 PAAR
       162,00 €
    Specifications:
    More
    Specifications
    Model
    Palm Fit 540D
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    HPPE
    Finish
    Coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4241B)