VWR® HistoChoice®, Tissue fixative with alcohol
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HistoChoice®
H112-30MLEA 9.2 EUR
H112-30ML
VWR® HistoChoice®, Tissue fixative with alcohol
Kleurstoffen Microscopy and staining reagents
HistoChoice® tssue fixative in pre-filled cups is a safe, convenient solution, to on-site tissue collection and transport. The wide mouth, pre-labelled cups are designed for use in doctors' offices, operating rooms, emergency rooms and outpatient clinics. HistoChoice® tissue fixative directly replaces common fixatives, including formalin-based, alcohol-based, Zenker's, B5, and Bouin's fixatives. Additionally, it can also be used in sequence with other fixatives. The preservative characteristics make it more effective than standard formaldehyde-based products. It yields vibrant staining characteristics and consistent results, even after long-term fixation. It preserves tissue integrity, eliminating the need for marker recovery procedures.
  • No reagent for recovery procedures as in formalin-based fixatives
  • Eliminates the need for ventilation systems and reduces disposal needs
  • Compatible with manual and automated processing procedures and equipment
This tissue fixative is suitable for use in histopathology, cytology, histochemistry, immunohistochemistry and molecular pathology.