Biopsy cassettes, ActivFlo™ Routine I
Supplier: LEICA HISTOLOGY
Certificates
About this item
ActivFlo Routine I cassettes are processing cassettes featuring a patented lateral vent design for maximum fluid exchange. The increase in fluid exchange results in less incidence of unfixed or under-processed tissues and faster fixation/processing of tissue specimens. These one piece cassettes have ‘frangible’ lids for easy removal. The cassettes are designed for optimal workflow in the Leica IP C Cassette Printer. They offer <1% printing error rate.
- With 45° writing surface
- Available in taped packaging for easy printer loading
- Eleven colours in quantities of 1000 available
Loose
Product Information
- Catalog #Supplier #ColourPkAvailabilityPrice
- LooseSupplier Article Number: 39LC-500-1Local Article Number: LEIH39LC-500-1Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-9Local Article Number: LEIH39LC-500-9Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-3Local Article Number: LEIH39LC-500-3Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-4Local Article Number: LEIH39LC-500-4Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-5Local Article Number: LEIH39LC-500-5Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-2Local Article Number: LEIH39LC-500-2Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-6Local Article Number: LEIH39LC-500-6Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-7Local Article Number: LEIH39LC-500-7Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-8Local Article Number: LEIH39LC-500-8Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-10Local Article Number: LEIH39LC-500-10Specifications:
Specifications
Environmentally Preferable - LooseSupplier Article Number: 39LC-500-11Local Article Number: LEIH39LC-500-11Specifications:
Specifications
Environmentally Preferable
Taped
Product Information
- Catalog #Supplier #ColourPkAvailabilityPrice
- TapedSupplier Article Number: 39LC-500-1-LLocal Article Number: LEIH39LC-500-1-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-4-LLocal Article Number: LEIH39LC-500-4-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-5-LLocal Article Number: LEIH39LC-500-5-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-10-LLocal Article Number: LEIH39LC-500-10-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-6-LLocal Article Number: LEIH39LC-500-6-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-8-LLocal Article Number: LEIH39LC-500-8-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-2-LLocal Article Number: LEIH39LC-500-2-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-11-LLocal Article Number: LEIH39LC-500-11-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-3-LLocal Article Number: LEIH39LC-500-3-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-9-LLocal Article Number: LEIH39LC-500-9-LSpecifications:
Specifications
Environmentally Preferable - TapedSupplier Article Number: 39LC-500-7-LLocal Article Number: LEIH39LC-500-7-LSpecifications:
Specifications
Environmentally Preferable