Netherlands
Order Entry
VWR®, DNA ladders
undefined
VWR®, DNA ladders
Supplier:  VWR Chemicals
Certificates
  •  
    Pk
  • PCR ladder, 100 to 3000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    PCR ladder, 100 to 3000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    733-2579
    1 KIT
    1 * 1 Kit
    1 * 1 Kit - 191,00 €
    191,00 €
    1 * 1 Kit
       191,00 €
  • Low range DNA ladder, 100 to 1000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    Low range DNA ladder, 100 to 1000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    733-2578
    1 KIT
    1 * 1 Kit
    1 * 1 Kit - 224,00 €
    224,00 €
    1 * 1 Kit
       224,00 €
  • High range DNA ladder, 200 to 12000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    High range DNA ladder, 200 to 12000 bp, sufficient for 250 lanes (loading 2 μl per gel), 1×0,5 ml
    733-2577
    1 KIT
    1 * 1 Kit
    1 * 1 Kit - 270,00 €
    270,00 €
    1 * 1 Kit
       270,00 €