Fluidised bath, SB range
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TECNF932DEA 4346.32 EUR
TECNF932D TECNF946D
Fluidised bath, SB range
Baden Gefluïdiseerde baden
The SB range of fluidised baths offers a safer alternative to the dangers associated with high temperature oil and salt baths. These units are designed to be bench or floor standing and only require an electrical and air supply for operation. Air passes through the mass of the aluminium oxide (Alundum) particles via a porous plate in the base of the unit separating the individual particles and suspending them in free air, giving the properties of a liquid bath.
Heaters are fitted within the bath which allow temperatures of 500 to 600 °C to be maintained. Both models in the SB range feature a stainless-steel inner container insulated from the outer wall, a safety air pressure switch to protect against loss of air pressure and also a thermal cut-out. It is highly recommended that the TC-9D temperature control unit is used in conjunction with these baths, greatly improving temperature stability and accuracy.
Leveringsinformatie: SBL-2D 230 V 50 to 60 Hz supplied with 50 kg alundum and overspill flange.