Netherlands
Order Entry
Temperature data loggers
Temperature data loggers
Supplier:  COMARK
Certificates
Temperature data loggers
Supplier:  COMARK
Specifications
  • Catalog No:
  • COMAN2013
  • COMAN2011
  • COMAN2012
  • COMAN2014
  • Sensor:
  • T: Thermistor
    RH: Capacitive
  • Thermistor
  • Thermistor
  • Int.: Thermistor
    Ext.: Types T/K thermocouple
  • Display:
  • LCD
  • LCD
  • LCD
  • LCD
  • Battery life:
  • 5 years
  • 5 years
  • 5 years
  • 5 years
  • Recording intervals:
  • 1 s - 99 hr
  • 1 s - 99 hr
  • 1 s - 99 hr
  • 2 s - 99 hr
  • Response time:
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • W×D×H:
  • 88×80×35 mm
  • 88×80×35 mm
  • 88×80×35 mm
  • 88×80×35 mm
  • Data memory - No. of readings:
  • 16000
  • 16000
  • 16000
  • 16000
  • Batteries:
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
Specifications
Product Information
  • Catalog #
    Supplier #
    Range
    Accuracy
    Resolution
    Pk
    Availability
    Price
  • Temperature data logger with internal thermistor
    Temperature data logger with internal thermistor
    COMAN2011
    N2011
    T: –40…+70 °C
    T: ±0,5 °C (–25...+50 °C); ±1,0 °C (–40...+70 °C)
    T: 0,1 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 464,00 €
    464,00 €
    1 * 1 ST
       464,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Sensor
    Thermistor
    Display
    LCD
    Interface
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • Temperature data logger with one internal and four external temperature sensors
    Temperature data logger with one internal and four external temperature sensors
    COMAN2012
    N2012
    T: –40…+70 °C Int.;
    T: –40…+150 °C Ext.
    T: ±0,5 °C (–25…+50 °C); ±1,0 °C (–40…+70 °C); ±2,0 °C(70...150 °C)
    T: 0,1 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 597,00 €
    597,00 €
    1 * 1 ST
       597,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Sensor
    Thermistor
    Display
    LCD
    Interface
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • Single-use temperature logger, with humidity alarm
    Single-use temperature logger, with humidity alarm
    COMAN2013
    N2013
    T: –20…+60 °C
    RH: 0 - 97 %
    T: ±0,5 °C (–20...+50 °C); ±0,5 °C (50...60 °C); ±1 °C otherwise
    RH: ±3% (–20...+60 °C)
    T: 0,1 °C
    RH: 0.1 %
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 847,00 €
    847,00 €
    1 * 1 ST
       847,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Sensor
    T: Thermistor
    RH: Capacitive
    Display
    LCD
    Interface
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • Temperature data logger, single/multiple thermocouple
    Temperature data logger, single/multiple thermocouple
    COMAN2014
    N2014
    T: –200…+1372 °C (K type)
    T: –200…+400 °C (T type)
    T: –40…+70 °C (thermistor)
    T: ±0,5 °C (–20…+40 °C); ±1,0 °C (–40…+70 °C); ±0,5 °C at 20 °C; ±0,3 °C (–20…+40 °C, thermistor); ±0.5 °C (–40…+70 °C, thermistor)
    0,2 °C (0…300 °C); 0,5 °C (>300 °C)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 641,00 €
    641,00 €
    1 * 1 ST
       641,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Sensor
    Int.: Thermistor
    Ext.: Types T/K thermocouple
    Display
    LCD
    Interface
    Battery life
    5 years
    Recording intervals
    2 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery