Netherlands
Order Entry
Petri dishes, standard
Petri dishes, standard
Supplier:  BOETTGER PAUL
Certificates
undefined
Petri dishes, standard
Supplier:  BOETTGER PAUL
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Version
    Ø
    Height
    Sterility
    Packed
    Pk
    Availability
    Price
  • BOET04.031.2100
    04.031.2100
    With vents, clear
    94 mm
    16 mm
    Sterile
    20/bag
    480
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 114,00 €
    114,00 €
    1 * 480 ST
       114,00 €
    Supplier Article Number: 04.031.2100
    Local Article Number: BOET04.031.2100
    Specifications:
    Specifications
    Environmentally Preferable
  • BOET04.041.2100
    04.041.2100
    Without vents, clear
    94 mm
    16 mm
    Sterile
    20/bag
    480
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 114,00 €
    114,00 €
    1 * 480 ST
       114,00 €
    Supplier Article Number: 04.041.2100
    Local Article Number: BOET04.041.2100
    Specifications:
    Specifications
    Environmentally Preferable