These non woven wipes made of viscose rayon are ideal for the disinfection of hard surfaces. Isopropyl alcohol, the active component of Azowipe™, is acknowledged by leading authorities to be an effective medium for the routine disinfecting of clinical surfaces. It kills bacteria in under a minute - proven to be effective against a variety of organisms including MRSA.
- Convenient for use in all clinical environments
- Gamma irradiated, sealed and sterilised in double polythene bags; with irradiation dot
- Specially designed canister for easy dispensing
- Foil sealed - prevents evaporation in storage
- Contain no preservatives - no smearing or streaking when in use
Verpakking: 200 wipes per tub dispenser. Each tub dispenser is double bagged and affixed with a Gamma Ray Indicator Dot.
Opgelet: Use biocides safely. Always read the label and product information before use.
*De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904