Power supply, programmable, Owl™ EC-3000XL2
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OWLAEC3000XL2EA 4120 EUR
OWLAEC3000XL2
Power supply, programmable, Owl™ EC-3000XL2
Ideal for DNA, RNA and protein electrophoresis, the Owl™ EC-3000XL2 power supply has a power-off memory that retains settings after shut-down. Owl™ EC-3000XL2 power supply is simple to use, multi-purpose and lightweight. Includes soft-touch key pad and three jacks.