Fluorescent ruler, SP Bel-Art
Leverancier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP Bel-Art
BELAF133550001EA 48.8 EUR
BELAF133550001
Fluorescent ruler, SP Bel-Art
Linialen
Fluorescent ruler can be used in observing and photographing ethidium bromide stained nucleic acid agarose and acrylamide gels.
  • Ruler fluorescences visibly upon illumination with ultraviolet light
  • Measure of nucleic acid mobility or quantify the size of nucleic acid fragments
  • Measurement allows for comparison of gels run at different times under the same conditions