/
Netherlands
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Digital clock/timer
COMAUTL882
:
COMARK
Digital clock/timer
COMAUTL882
:
COMARK
103,00 €
1 * 1 ST
-
+
Colour:
White/grey
Description:
Digital clock/timer, capacity of 24 h
Environmentally Preferable:
Pk:
1