Flow meter, float type, Accutek®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENTEMFM-6N-150-PEA 0 EUR
ENTEMFM-6N-150-P ENTEFM-8F-13250-P
Flow meter, float type, Accutek®
Accutek flow meters are designed for precise metering of corrosive and/or ultrapure fluids and to minimise entrapment areas.
  • All wetted parts on the flowmeter are constructed of PFA, PTFE and CTFE
  • Integral Flaretek® tube fitting and FNPT ports are standard
  • Sight tubes are molded from PFA HP Plus material, providing better clarity, increased visibility and a cleaner flow path
  • Sight tubes come standard with easy to read black laser marked numbers
  • Performance is calibrated (in water) for an accuracy of ±5,0 % of full scale