Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Cloning cylinders
Leverancier: HILGENBERG GMBH
Borosilicate glass 3.3.
  • High-grade borosilicate glass rings
  • Thermally sterilized
  • Wall thickness: 1 mm
Cloning cylinders or rings with ground edges are used to seperate the individual cells or cell groups in the nutrient soultion by placing the ring over it.The ring requires little grease to provide an optimum seal between the ring and the petri dish.
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
Hoogte Ø uitw. VPE VWR catalogus nummer Hoeveelheid
8 mm 6 mm 100 HILG1980004
8 mm 8 mm 1 710-0203
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid